missing translation for 'onlineSavingsMsg'
Learn More

FBXO7 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marke:  Proteintech 10696-1-AP-150UL

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16887243

  • 329.85 € / 150 Mikroliter

Verfügbar ab
16-06-2021

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbxs class and it may play a role in regulation of hematopoiesis. Alternatively spliced transcript variants of this gene have been identified with the full-length natures of only some variants being determined.
Spezifikation

Spezifikation

FBXO7
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q3U7U3, Q9Y3I1
F box only protein 7, F box protein 7, FBX, FBX07, FBX7, FBXO7, PARK15, PKPS
Rabbit
IgG
150 μL
-20°C
Primary
25793, 69754
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
FBXO7
Liquid
Fbxo7
FBXO7 Fusion Protein Ag1035
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation