missing translation for 'onlineSavingsMsg'
Learn More

FCGR3A, 176 Phe/Val Recombinant Human Protein, His Tag

FCGR3A, 176 Phe/Val Recombinant Human Protein, His Tag

Marke:  Invitrogen 10389H08C1250

Weitere Details : Netto-Gewicht : 0.01000kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13412999

  • 864.00 € / 250 Mikrogramm


Voraussichtlicher Liefertermin 05-04-2019
Zum Warenkorb hinzufügen

For Research Use Only. All usage must comply with product instructions.


Beschreibung

Beschreibung

Fc fragment of IgG, low affinity IIIa, receptor (FCGR3A/CD16a), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1- Gln 208) of human FCGR3A (P08637-1), (176 Phe/Val), to fused to a polyhistidine tag at the C-terminus.

Activity: This recombinant protein has not been tested.

Formulation: Lyophilized in 140 mM NaCl, 2.7 mM KCl, 10 mM Na2HPO4, 1.8 mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.



This protein is manufactured by Sino Biological Inc.

Spezifikation

Spezifikation

Protein assays and analysis
Lyophilized
FCGR3A 176 Phe/Val Recombinant Human Protein
5 x 50μg
For research use only. Not for use in diagnostic procedures
Other proteins
FCGR3A 176 Phe/Val Recombinant Human Protein
FCGR3A
CD proteins, growth factors and receptors
FCGR3A 176 Phe/Val Recombinant Human Protein
His Tag
Lyophilized
23.3kDa
≥95% by SDS-PAGE
–20°C
5 Pack(s)
RP11-5K23.1, CD16, CD16A, FCG3, FCGR3, FCGRIII, FCR-10, FCRIII, FCRIIIA, IGFR3
5 Vials of lyophilized protein
Assay standard
Mammalian
Human
Videos

For Research Use Only. Not for use in diagnostic procedures.