missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ SARS-CoV-2 Nucleocapsid ELISA Kit (Colorimetric)

Product Code. 18602630 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18602630 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18602630 Supplier Novus Biologicals™ Supplier No. NBP311405
 Restricted Item
Verfügbar ab: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Recognizes both recombinant and natural SARS-CoV-2 Coronavirus Nucleocapsid

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 93.75-6000 pg/mL
Assay Sensitivity 46.68 pg/mL
Product Type ELISA Kit (Colorimetric)
Sample Type Cell Culture Supernatant, Serum
Specificity Recognizes both recombinant and natural SARS-CoV-2 Coronavirus Nucleocapsid
For Use With (Application) ELISA, Sandwich ELISA
Gene ID (Entrez) 43740575
Gene Symbol N
Quantity 1 Kit
Research Discipline Virology Bacteria and Parasites
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym 2019-nCoV N Protein, 2019-nCoV Nucleocapsid Protein, COVID-19 N protein, COVID-19 nucleocapsid protein, Human coronavirus Nucleoprotein, nucleocapsid phosphoprotein, Nucleocapsid protein, Nucleoprotein, ORF9; structural protein, SARS-CoV-2, SARS-CoV-2 Nucleocapsid protein, SARS-CoV-2 N protein, SARSCoV2 Nucleoprotein, Severe Acute Respiratory Syndrome Coronavirus 2 Nucleocapsid Protein
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.