Learn More
MP Biomedicals™ Fetuin, From Fetal Bovine Serum
Fetuin is a protein containing 12 Cys residues, all involved in disulfide bridges.
Marke: MP Biomedicals™ 0210487401
Weitere Details : CAS : 9014-81-7 Netto-Gewicht : 0.01000kg
Beschreibung
- Formulation: Polypeptide 74%, Hexose 8.3%, Hexosamines 5.5%, and Sialic Acid 8.7%
Applications:
Fetuin bovine is a Protein with the ability to bind to the insulin receptor and inhibit insulin action, Fetuin has been used in studies of type 2 diabetes mellitus (T2DM), In patients with T2DM, fetuin has been shown to be significantly lower when compared to control patients.
Spezifikation
Type 2 Diabetes Mellitus (T2DM) Studies, Cell Attachment and Proliferation | |
Fetuin Protein | |
1g | |
Powder |
Polypeptide 74%, Hexose 8.3%, Hexosamines 5.5%, and Sialic Acid 8.7% | |
Ammonium Sulfate Fractionation | |
Fetal Bovine Serum |