Learn More
Invitrogen™ FGF2 Human ELISA Entwicklerkit (ABTS ), PeproTech® PeproTech®
Artikelnummer. 17848995
Change view
Click to view available options
Menge:
2 x 96 Tests
Packungsgröße:
2 Stück
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
17848995 2 x 96 Tests 2 Stück
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 17848995 Lieferant Invitrogen™ Lieferanten-Nr. 900M08
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
ELISA
FGF2 (FGFb, fibroblast growth factor basic) belongs to the fibroblast growth factor (FGF) family, and interacts with high-affinity transmembrane receptors to influence cell proliferation and tissue neovascularization. FGF2 exists as five isoforms with distinct intracellular localizations and functions. The 18 kDa isoform is predominantly cytosolic and acts through cell surface receptors, whereas the 22, 22.5, 24 and 34 kDa isoforms are nuclear and may signal independent of transmembrane receptor pathways. In humans, the gene is located on the q arm of chromosome 4. FGF2 has been implicated in diverse biological processes, such as limb and nervous system development, wound healing, and tumor growth. The mRNA for FGF2 contains multiple polyadenylation sites, and is alternatively translated from non-AUG and AUG initiation codons, resulting in five different isoforms with distinct properties. The CUG-initiated isoforms are localized in the nucleus and are responsible for the intracrine effect, whereas, the AUG-initiated form is mostly cytosolic and is responsible for the paracrine and autocrine effects of this FGF. Diseases associated with FGF2 dysfunction include Kaposi Sarcoma and corneal neovascularization.
TRUSTED_SUSTAINABILITY
Spezifikation
Zugriffsnummer P09038
Produkttyp ELISA
Gen-ID (Entrez) 2247
Gensymbol FGF2
Menge 2 x 96 Tests
Kennzeichnung RUO
Lagerungsbedingungen Informationen zur komponentenspezifischen Lagertemperatur finden Sie in der Produktdokumentation.
Ziel Human