missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGF9 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 26554-1-AP-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. This protein was isolated as a secreted factor that exhibits a growth-stimulating effect on cultured glial cells. In nervous system, this protein is produced mainly by neurons and may be important for glial cell development. Expression of the mouse homolog of this gene was found to be dependent on Sonic hedgehog (Shh) signaling. Mice lacking the homolog gene displayed a male-to-female sex reversal phenotype, which suggested a role in testicular embryogenesis.Spezifikation
FGF9 | |
Polyclonal | |
Unconjugated | |
FGF9 | |
FGF 9, FGF9, Fibroblast growth factor 9, GAF, Glia activating factor, HBFG 9, HBGF 9 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
2254 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry | |
0.47 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P31371 | |
FGF9 | |
FGF9 Fusion Protein Ag24244 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |