missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGF9 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00 € - 399.00 €
Spezifikation
Antigen | FGF9 |
---|---|
Konzentration | 0.47 mg/mL |
Anwendungen | Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry |
Klassifikation | Polyclonal |
Konjugat | Unconjugated |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16884975
|
Proteintech
26554-1-AP-20UL |
20 μL |
140.00 €
20 Mikroliter |
Verfügbar ab: 23-04-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
16874975
|
Proteintech
26554-1-AP-150UL |
150 μL |
399.00 €
150 Mikroliter |
Verfügbar ab: 23-04-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. This protein was isolated as a secreted factor that exhibits a growth-stimulating effect on cultured glial cells. In nervous system, this protein is produced mainly by neurons and may be important for glial cell development. Expression of the mouse homolog of this gene was found to be dependent on Sonic hedgehog (Shh) signaling. Mice lacking the homolog gene displayed a male-to-female sex reversal phenotype, which suggested a role in testicular embryogenesis.Spezifikation
FGF9 | |
Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Human | |
P31371 | |
2254 | |
FGF9 Fusion Protein Ag24244 | |
Primary | |
-20°C | |
FGF9 |
0.47 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
FGF 9, FGF9, Fibroblast growth factor 9, GAF, Glia activating factor, HBFG 9, HBGF 9 | |
FGF9 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |