missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ FGL1 (Human) Recombinant Protein
Human FGL1 full-length ORF ( AAH07047, 19 a.a. - 312 a.a.) recombinant protein with GST-tag at N-terminal.
Marke: Abnova™ H00002267-P01.25ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
- Sequence: EISALEDCAQEQMRLRAQVRLLETRVKQQQVKIKQLLQENEVQFLDKGDENTVIDLGSKRQYADCSEIFNDGYKLSGFYKIKPLQSPAEFSVYCDMSDGGGWTVIQRRSDGSENFNRGWKDYENGFGNFVQKHGEYWLGNKNLHFLTTQEDYTLKIDLADFEKNSRYAQYKNFKVGDEKNFYELNIGEYSGTAGDSLAGNFHPEVQWWASHQRMKFSTWDRDHDNYEGNCAEEDQSGWWFNRCHSANLNGVYYSGPYTAKTDNGIVWYTWHGWWYSLKSVVMKIRPNDFIPNVI
Spezifikation
AAH07047.1 | |
fibrinogen-like 1 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FGL1 | |
GST |
2267 | |
Wheat germ expression system | |
25 μg | |
HFREP1, HP-041, LFIRE1, MGC12455 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |