Learn More
Thermo Scientific™ FH 40 TG Telescopic Probes
Product Code. 17898690 Shop All Thermo Scientific Products
Change view
Click to view available options
Energy Range:
60 keV to 1.3 MeV
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Energy Range Unit Size
17898690 60 keV to 1.3 MeV Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17898690 Supplier Thermo Scientific™ Supplier No. 425405310
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here
missing translation for 'validMainframeMsg'
Measure dose and dose rate at a secure distance for all situations where an extended range is required or high radiation may occur.
Measure dose and dose rate at a secure distance for all situations where an extended range is required or high radiation may occur with the Thermo Scientific™ FH 40 TG Telescopic Probe, which attaches to the Thermo Scientific™ FH 40 GL Digital Survey Meter.
  • Simultaneously determines dose and dose rate measurement at the operator end while measuring dose rate at the teleprobe end
  • Features a built-in speaker and the ability to log up to 256 measurements for later retrieval by a PC via infrared serial interface
  • A large backlit digital display with an analog graph and menu-driven operator functions facilitate a user-friendly interface
  • Modular design allows easy exchange of components
  • Extendible up to 4m (approx. 13ft.)
  • Reinforced, isolating fiberglass material, fully watertight
  • Half the weight of conventional teledetectors
  • Interchangeable probes for different measurement tasks
  • “Smart probes” with individual calibration
  • Standard detector FHZ 612 contains two energy-compensated GM tubes
  • Acoustic alarm levels can be set independently for both detectors
  • Options include 1 × 1in. NaI detector head for high gamma sensitivity, neutron “sniffer” detector head, and beta-sensitive probe; options also include aluminum case and FH 40 G-X
  • Display-Unit (FH 40 G without internal detector)
Recommended for:
  • Nuclear power plants
  • Nuclear industry
  • Recycling industry
  • Emergency response
  • Fire brigades
  • Nuclear medicine
  • Research
TRUSTED_SUSTAINABILITY
Specifications
Description FH 40 TG-10; Enrgy range: 60 keV - 1.3 MeV
Energy Range 60 keV to 1.3 MeV
Detector Type Two energy compensated GM tubes (low and high range)
Height (English) 38 in.
Height (Metric) 950 mm
Internal Detector Proportional tube (supplied with Model TeleProbe40GL only) 10 µR/h to 10 R/h (100 nSv/h to 100 mSv/h)
Depth (English) 3.2 in.
Depth (Metric) 80 mm
Material Reinforced, isolating fiberglass material, fully watertight
Measuring Range 0.01 µSv/h to 10 Sv/h
Weight (English) 6.6 lb.
Weight (Metric) 3 kg
Width (English) 2 in.
Width (Metric) 52 mm
Radiation Type Dependent on probe used
Product Type Telescopic Probe