missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ FHL3 (Human) Recombinant Protein
Human FHL3 full-length ORF ( AAH01351.1, 1 a.a. - 280 a.a.) recombinant protein with GST-tag at N-terminal.
Marke: Abnova™ H00002275-P01.25ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
- Sequence: MSESFDCAKCNESLYGRKYIQTDSGPYCVPCYDNTFANTCAECQQLIGHDSRELFYEDRHFHEGCFRCCRCQRSLADEPFTCQDSELLCNDCYCSAFSSQCSACGETVMPGSRKLEYGGQTWHEHCFLCSGCEQPLGSRSFVPDKGAHYCVPCYENKFAPRCARCSKTLTQGGVTYRDQPWHRECLVCTGCQTPLAGQQFTSRDEDPYCVACFGELFAPKCSSCKRPIVGLGGGKYVSFEDRHWHHNCFSCARCSTSLVGQGFVPDGDQVLCQGCSQAGP
Spezifikation
AAH01351.1 | |
four and a half LIM domains 3 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FHL3 | |
GST |
2275 | |
Wheat germ expression system | |
25 μg | |
MGC19547, MGC23614, MGC8696, SLIM2 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |