missing translation for 'onlineSavingsMsg'
Learn More

Filensin Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marke:  Proteintech 17492-1-AP-150UL

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16887384

  • 329.85 € / 150 Mikroliter

Voraussichtliches Versanddatum: 25-06-2021

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

More than 99% of the vertebrate ocular lens is comprised of terminally differentiated lens fiber cells. Two lens-specific intermediate filament-like proteins, CP49 (also known as phakinin) and the protein product of this gene, filensin, are expressed only after fiber cell differentiation has begun. Both proteins are found in a structurally unique cytoskeletal element that is referred to as the beaded filament (BF). Mutations in this gene are the cause of autosomal recessive cortical juvenile-onset cataract. Multiple transcript variants encoding different isoforms have been found for this gene.
Spezifikation

Spezifikation

Filensin
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q12934
BFSP1, CP115, CP94, Filensin, LIFL H
Rabbit
IgG
150 μL
-20°C
Primary
631
Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
BFSP1
Liquid
BFSP1
Filensin Fusion Protein Ag11591
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Zertifikate