missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FKBP5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 14155-1-AP-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
FKBP5 is a member of the immunophilin protein family. This family plays a role in immunoregulation and basic cellular processes involving protein folding and trafficking. FKBP5 is a cis-trans prolyl isomerase that binds to the immunosuppressants FK506 and rapamycin. It is thought to mediate calcineurin inhibition. It also interacts functionally with mature hetero-oligomeric progesterone receptor complexes along with the 90 kDa heat shock protein and P23 protein. The gene that encodes FKBP5 has been found to have multiple polyadenylation sites. Alternative splicing results in multiple transcript variants.Spezifikation
FKBP5 | |
Polyclonal | |
Unconjugated | |
FKBP5 | |
51 kDa FK506 binding protein, 51 kDa FKBP, AIG6, Androgen regulated protein 6, FF1 antigen, FK506 binding protein 5, FKBP 5, FKBP 51, FKBP5, FKBP51, FKBP54, HSP90 binding immunophilin, P54, PPIase, PPIase FKBP5, Ptg 10, Rotamase | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14229, 2289, 361810 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.26 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q13451, Q64378 | |
Fkbp5 | |
FKBP5 Fusion Protein Ag5337 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |