Learn More
Abnova™ FKBP8 Recombinant Protein
Human FKBP8 full-length ORF ( AAH09966, 1 a.a. - 355 a.a.) recombinant protein with GST-tag at N-terminal
Marke: Abnova™ H00023770-P01.25ug
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
The protein encoded by this gene is a member of the immunophilin protein family, which play a role in immunoregulation and basic cellular processes involving protein folding and trafficking. Unlike the other members of the family, this encoded protein does not seem to have PPIase/rotamase activity. It may have a role in neurons associated with memory function. (provided by RefSeq)
- Molecular weight: 64.79kDa
- Preparation method: in vitro wheat germ expression system
- Purification: Glutathione Sepharose 4 Fast Flow
- Storage Buffer:50mM Tris-HCI, 10mM reduced Glutathione, pH 8 in the elution buffer
- Quality Control Testing: 12.5% SDS-PAGE stained with Coomassie Blue
Best use within three months from the date of receipt of this protein
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Spezifikation
AAH09966 | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
64.79 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
MGQPPAEEAEQPGALAREFLAAMEPEPAPAPAPEEWLDILGNGLLRKKTLVPGPPGSSRPVKGPVVTVHLQTSLENGTRVQEEPELVFTLGDCDVIQALDLSVPLMDVGETAMVTADSKYCYGPQGRSPYIPPHAALCLEVTLKTAVDGPDLEMLTGQERVALANRRRECGNAHYQRADFVLAANSYDLAIKAITSSAKVDMTFEEEAQLLQLKVKCLNNLAASQLKLDHYRAALRSCSLVLEHQPDNIKALFRKGKVLAQQGEYSEAIPILRAALKLEPSNKTIHAELSKLVKKHAAQRSTETALYRKMLGNPSRLPAKCPGKGAWSIPWKWLFGATAVALGGVALSVVIAARN | |
FKBP38/FKBPr38 | |
FKBP8 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Protein Array, ELISA, Western Blot | |
23770 | |
FKBP8 (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FKBP8 | |
Human | |
Recombinant | |
Solution |