Learn More
FKBPL Mouse anti-Human, Mouse, Rat, Clone: 2H10B10, Proteintech
Artikelnummer. 16828225 Alle ansehen Proteintech Produkte
Change view
Click to view available options
Menge:
20 μL
150 μL
Packungsgröße:
150 Mikroliter
20 Mikroliter
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
16828225 150 μL 150 Mikroliter
16838225 20 μL 20 Mikroliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 16828225 Lieferant Proteintech Lieferanten-Nr. 663891IG150UL
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Mouse Monoclonal Antibody
The protein encoded by this gene has similarity to the immunophilin protein family, which play a role in immunoregulation and basic cellular processes involving protein folding and trafficking. The encoded protein is thought to have a potential role in the induced radioresistance. Also it appears to have some involvement in the control of the cell cycle.
Spezifikation
Antigen FKBPL
Anwendungen Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Klassifikation Monoclonal
Klon 2H10B10
Konzentration 1 mg/mL
Konjugat Unconjugated
Zusammensetzung PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gen FKBPL
Gen-Zugriffsnummer O35450, Q6MG81, Q9UIM3
Gen-Alias DIR1, FK506 binding protein like, FKBPL, NG7, WISP39
Gensymbole Fkbpl
Wirtsspezies Mouse
Immunogen FKBPL Fusion Protein Ag0112
Reinigungsverfahren Protein G
Menge 150 μL
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 406168, 56299, 63943
Zielspezies Human, Mouse, Rat
Inhalt und Lagerung -20°C
Produkttyp Antibody
Form Liquid
Isotype IgG1