Learn More
Idex X10 FITTINGS 1/16 für Acetal PR Schlauch
No need to flange tubing with the Flangeless Ferrule.
Marke: Idex 01939-30
Weitere Details : Netto-Gewicht : 0.80000kg
Beschreibung
- Excellent for connecting rigid or semi-rigid tubing.
- Use with male nuts for a leak-tight connection.
Spezifikation
Flanschlose Fittings | |
1,5875 mm | |
10/Packung |
0.0625 in. | |
0193930 | |
25,4 |