missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Fluorometric and Luminometric Filter Sets for Varioskan™ ALF Multimode Microplate Reader

Artikelnummer. 30178795 Alle ansehen Thermo Scientific Produkte
Ansicht ändern
Click to view available options
Anregung/Emission:
334/425 nm
334/450 nm
390/450 nm
390/513 nm
390 nm, 450/525 nm (Spender/Akzeptor)
438/497 nm
438 nm, 497/542 nm (Spender/Akzeptor)
452/545 nm
460 nm, 618 nm (Spender/Akzeptor)
485/575 nm
485 nm, 525/595 nm (Spender/Akzeptor)
497/542 nm
527/615 nm
535/595 nm
535 nm, 595/677 nm (Spender/Akzeptor)
555/605 nm
625/677 nm
649/711 nm
Packungsgröße:
1 Stück
18 product options available for selection
Product selection table with 18 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Anregung/Emission Packungsgröße
30178795 460 nm, 618 nm (Spender/Akzeptor) 1 Stück
30178788 452/545 nm 1 Stück
30178789 497/542 nm 1 Stück
30178782 535/595 nm 1 Stück
30178794 555/605 nm 1 Stück
30178798 625/677 nm 1 Stück
30178784 649/711 nm 1 Stück
30178791 390/513 nm 1 Stück
30178786 390/450 nm 1 Stück
30178787 334/450 nm 1 Stück
30178796 438/497 nm 1 Stück
30178792 334/425 nm 1 Stück
30178793 527/615 nm 1 Stück
30178783 485/575 nm 1 Stück
30178781 438 nm, 497/542 nm (Spender/Akzeptor) 1 Stück
30178799 485 nm, 525/595 nm (Spender/Akzeptor) 1 Stück
30178785 535 nm, 595/677 nm (Spender/Akzeptor) 1 Stück
30178797 390 nm, 450/525 nm (Spender/Akzeptor) 1 Stück
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
18 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 30178795 Lieferant Thermo Scientific™ Lieferanten-Nr. VAPAI006
  • ONLINE EXCLUSIVE PRICE
    936.00 €
    786.24 € / 1 Stück
    Sparen 149.76 €
    16% Rabatt
Alle Werbeaktionen anzeigen
 Eingeschränkter Artikel
Verfügbar ab: 21-10-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Varioskan ALF Multimode Microplate Reader Filter Sets are high quality, validated, and available for various fluorometric and luminometric applications.

With the Varioskan ALF Multimode Microplate Reader, fluorescence and certain luminescence measurements are performed with filter-based optics for optimal specificity and sensitivity. The system comes with three pre-installed excitation and emission filter sets suitable for many common fluorometric applications like Quant-iT DNA quantitation assays and PrestoBlue and AlamarBlue cell viability assays. Additionally, five open filter pair positions are available for customized fluorometric and luminometric applications such as measurement of various Alexa Fluor™ and other common dyes or Promega NanoBRET™ assays. The filters are available as predefined, carefully tested, and validated filter sets, to ensure the optimal performance of selected assays.

Varioskan ALF instrument includes the following factory installed fluorometric filter pairs:

  • Coumarin family filter pair
    • Excitation 345 nm, emission 450 nm
  • Fluorescein family filter pair
    • Excitation 485, emission 525 nm
  • Resorufin family filter pair
    • Excitation 555 nm, emission 615 nm

In addition to factory installed filters, a range of validated filter sets are available for various fluorometric and luminometric applications.

  • Filter sets consists of high transmission filters for high excitation and emission efficiency.
  • Filter sets are designed to efficiently block unwanted light as well as unwanted emitted light to ensure high signal and low background.
  • Varioskan ALF filter installation is effortless
    • The instrument filter wheel is easy to access, and the correct parameters are quickly set in simple workflow in Thermo Scientific SkanIt software.
    • SkanIt software-controlled filter installation process prevents accidental mistakes ensuring error-free
TRUSTED_SUSTAINABILITY

Spezifikation

Produkttyp Filter Set
Zur Verwendung mit (Geräte) Varioskan ALF Multimode Microplate Reader
Plattentyp 6 to 384-well microplates
Anregung/Emission 460 nm, 618 nm (Spender/Akzeptor)
Menge Each
Zur Verwendung mit Promega NanoBRET™ assays

Zur Verwendung im Labor

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.