missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FLVCR1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 26841-1-AP-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
This gene encodes a member of the major facilitator superfamily of transporter proteins. The encoded protein is a heme transporter that may play a critical role in erythropoiesis by protecting developing erythroid cells from heme toxicity. This gene may play a role in posterior column ataxia with retinitis pigmentosa and the hematological disorder Diamond-Blackfan syndrome.Spezifikation
FLVCR1 | |
Polyclonal | |
Unconjugated | |
FLVCR1 | |
FLVCR, FLVCR1, hFLVCR | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
226844, 28982 | |
-20°C | |
Liquid |
Western Blot, Immunofluorescence, Immunocytochemistry, Immunoprecipitation | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
B2RXV4, Q9Y5Y0 | |
FLVCR1 | |
FLVCR1 Fusion Protein Ag25220 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |