missing translation for 'onlineSavingsMsg'
Learn More

FMO4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marke:  Proteintech 15394-1-AP-150UL

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16884544

  • 329.85 € / 150 Mikroliter

Voraussichtliches Versanddatum: 18-06-2021

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

This antibody, generated by N-330 amino-acids of FMO4, can recognize the 2 variants and also cross-react with other FMO proteins.

FMO4 (Flavin-containing monooxygenase 4) also known as FMO2, FORMERLY or FMO, ADULT LIVER FORM, is an enzyme that in humans is encoded by the FMO4 gene. By PCR analysis, Dolphin et al. (1992) mapped the FMO4 gene to chromosome 1. Southern blot hybridization with single exon probes demonstrated that human FMO4 and FMO1 are, in each case, the product of a single gene. Lawton et al. (1994) proposed a reclassification of mammalian FMOs. Under this system, FMO2 described by Dolphin et al. (1992) is now designated FMO4.
Spezifikation

Spezifikation

FMO4
0.2 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P31512, Q8K4B7, Q8VHG0
Dimethylaniline oxidase 4, FMO 4, FMO2, FMO4
Rabbit
IgG
150 μL
-20°C
Primary
226564, 2329, 246247
Western Blot
Unconjugated
FMO4
Liquid
Fmo4
FMO4 Fusion Protein Ag7590
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Videos
SDS
Documents