missing translation for 'onlineSavingsMsg'
Learn More

Folinic acid calcium salt hydrate, 98%

Product Code. 15446658
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. Quantity unitSize
15446658 1 g 1g
15456658 250 mg 250mg
2 options
This item is not returnable. View return policy

Product Code. 15446658

Brand: Thermo Scientific Alfa Aesar H60181.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1492-18-8 | C20H21CaN7O7 | 511.51 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1492-18-8
Molecular Formula C20H21CaN7O7
Molecular Weight (g/mol) 511.51
MDL Number MFCD00006704
InChI Key KVUAALJSMIVURS-AEQORCIRNA-L
Synonym calcium folinate
PubChem CID 131675590
IUPAC Name calcium;(4S)-4-[[4-[(2-amino-5-formyl-4-oxo-1,6,7,8-tetrahydropteridin-6-yl)methylamino]benzoyl]amino]-5-hydroxy-5-oxopentanoate
SMILES [Ca++].NC1=NC(=O)C2=C(NCC(CNC3=CC=C(C=C3)C(=O)N[C@@H](CCC([O-])=O)C([O-])=O)N2C=O)N1

Specifications

Quantity 1 g
Beilstein 5723924
Formula Weight 511.51 (Anhydrous)
Percent Purity 98%
Chemical Name or Material Folinic acid calcium salt hydrate
compliance-icons
Product Identifier
  • Folinic acid calcium salt hydrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Folinic acid calciumsalt hydrate, Calcium folinate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.