Learn More
Fomblin YR-1800, Thermo Scientific Chemicals
Catalog Number p-7047079
Click to view available options
Menge:
5 g
25 g
Regulatorischer Hinweis: Für den Kauf dieses Produkts gelten möglicherweise Einschränkungen. Etwaige zusätzliche Unterlagen werden bei Bedarf von Ihnen angefordert.
69991-67-9
C3F6
150.023
MFCD01323324
HCDGVLDPFQMKDK-UHFFFAOYSA-N
hexafluoropropylene, hexafluoropropene, perfluoropropene, perfluoropropylene, propene, hexafluoro, perfluoro-1-propene, propylene, hexafluoro, hexfluoropropylene, 1-propene, 1,1,2,3,3,3-hexafluoro, 1,1,2,3,3,3-hexafluoro-1-propene
8302
1,1,2,3,3,3-hexafluoroprop-1-en
C(=C(F)F)(C(F)(F)F)F
Regulatorischer Hinweis: Für den Kauf dieses Produkts gelten möglicherweise Einschränkungen. Etwaige zusätzliche Unterlagen werden bei Bedarf von Ihnen angefordert.
69991-67-9
C3F6
150.023
MFCD01323324
HCDGVLDPFQMKDK-UHFFFAOYSA-N
hexafluoropropylene, hexafluoropropene, perfluoropropene, perfluoropropylene, propene, hexafluoro, perfluoro-1-propene, propylene, hexafluoro, hexfluoropropylene, 1-propene, 1,1,2,3,3,3-hexafluoro, 1,1,2,3,3,3-hexafluoro-1-propene
8302
1,1,2,3,3,3-hexafluoroprop-1-en
C(=C(F)F)(C(F)(F)F)F
It is used in applications include pump and compressor cleaning, degreasing, dewatering, vapor blanketing, flash point reduction, and trace residue elimination. It acts as a diluent in the application of more viscous PFPE fluids and greases.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Fomblin YR-1800
Dichte 1.92
Löslichkeitsinformationen Difficult to mix in water.
Menge 5 g
Brechungsindex 1.304