missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Forma™ -20°C Enzyme Freezer

Artikelnummer. 17823318 Alle ansehen Thermo Scientific Produkte
Ansicht ändern
Click to view available options
Capacity (English):
23.3 cu. ft.
Plug Type:
UK
Voltage:
230 V
Packungsgröße:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Capacity (English) Plug Type Voltage Packungsgröße
17823318 23.3 cu. ft. UK 230 V Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 17823318 Lieferant Thermo Scientific™ Lieferanten-Nr. FFEN2320W

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Alternative Produkte anzeigen

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Maintain consistent temperatures with these manual defrost enzyme freezers, which feature gravity convection air systems without blowers or fans.

Maintain consistent temperatures with Thermo Scientific™ Forma™ -20°C Enzyme Freezers, which feature manual defrost and microprocessor control. Our Forma Enzyme Freezer is designed for storage applications where slight intermittent coil warming during an auto-defrost cycle is not desired. The gravity convection air system provides maximum stability without blowers or fans.

Microprocessor Control

  • Enhanced digital electronic technology simplifies operation and delivers greater accuracy
  • Operating temperature preset at -20°C
  • Icon-based controls and displays include graphic references
  • Setpoint security with key-operated triple-position master switch controls power to all systems
  • Audible/visual warnings for overtemperature, undertemperature and power failure
  • Visual status reports on critical functions
  • Graphic thermometer confirms operation: steady green for normal, flashing top bar for overtemperature, flashing bulb for undertemperature
  • Enlarged digital display with temperature resolution to within 0.1°C
  • Alarm silence, ringback and automatic reset functions
  • Pushbutton alarm test verifies sensor continuity
  • Battery backup
  • Remote alarm contacts

Refrigeration System

  • Heavy-duty, industrial-grade, hermetically sealed compressors
  • Manual defrost for maximum temperature uniformity performance

Construction

  • Industrial-quality cabinet construction
  • High-density, CFC-free urethane foam insulation
  • Solid door with keyed door lock
  • Door self-closes against closed-cell peripheral gasket
  • Full-length door handle
  • Adjustable leveling feet standard (optional casters available)
  • Scratch-resistant painted interior and exterior
  • Nine epoxy-coated, open-wire shelves, adjustable in 1″ (2.5cm) increments—no tools required

Includes:

  • 45 enzyme storage bins (5 per shelf); labels not provided
TRUSTED_SUSTAINABILITY

Spezifikation

Type Enzyme Freezer
Capacity (English) 23.3 cu. ft.
Capacity (Metric) 659 L
Temperature Range -20°C
Door Style Solid
No. of Doors 1
Cabinet Material Scratch-resistant Painted
Insulation Urethane Foam
Exterior Finish Epoxy Coated Steel
Shelves 9
Certifications/Compliance CE
Control Type Microprocessor
Warranty Warranty varies by country, please contact us for details
Defrost Manual
Dimensions (D x W x H) Interior 29 x 24 x 58 in. (73.7 x 61 x 147.3 cm)
Dimensions (L x W x H) Exterior 37.2 x 28 x 79.2 in. (94.5 x 71.1 x 201.2 cm)
Depth (English) Interior 29 in.
Depth (Metric) Interior 73.7 cm
Height (English) Exterior 79.2 in.
Height (English) Interior 58 in.
Height (Metric) Exterior 201.2 cm
Height (Metric) Interior 147.3 cm
Length (English) Exterior 37.2 in.
Length (Metric) Exterior 94.5 cm
Width (English) Exterior 28 in.
Width (English) Interior 24 in.
Width (Metric) Exterior 71.1 cm
Width (Metric) Interior 61 cm
Monitoring Options Alarm
Amperage 5.6 A
Breaker 15A
Plug Type UK
Voltage 230 V
Electrical Requirements 230 V 50 Hz
Frequency 50 Hz
Compressor Heavy-duty, Industrial-grade, Hermetic
Refrigerant CFC-free
Shipping Weight (English) 425 lb.
Shipping Weight (Metric) 193 kg
Rack Capacity 9
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.