missing translation for 'onlineSavingsMsg'
Learn More

Formaldehyde Solution, min. 37% GR for Analysis, ACS, Reagent Ph Eur Grade, MilliporeSigma™

Product Code. 10583531
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 L
Packaging:
Plastic bottle
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10583531 1 L Plastic bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10583531 Supplier Merck Chemicals Supplier No. 1040031000

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here

This item is not returnable. View return policy
For analysis

Chemical Identifiers

CAS 50-00-0 , 67-56-1
Molecular Formula CH2O
Molecular Weight (g/mol) 30.03
MDL Number MFCD00003274
InChI Key WSFSSNUMVMOOMR-UHFFFAOYSA-N
Synonym formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde
PubChem CID 712
ChEBI CHEBI:16842
IUPAC Name formaldehyde
SMILES C=O

Specifications

Ignition Point ca.300°C
Assay Percent Range min. 37%
Odor Stinging
Packaging Plastic bottle
Quantity 1 L
UN Number UN2209
Grade ACS/Reagent Ph. Eur.
Physical Form Liquid
Chemical Name or Material Formaldehyde solution, Stabilized with about 10% methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.