missing translation for 'onlineSavingsMsg'
Learn More

Ameisensäure, ACS, 96+%, Thermo Scientific Chemicals

Catalog Number p-7050370
Click to view available options
Menge:
500g
2kg
4 x 500g
64-18-6
CH2O2
46.025
MFCD00003297
BDAGIHXWWSANSR-UHFFFAOYSA-N
methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt
284
CHEBI:30751
Ameisensäure
C(=O)O
64-18-6
CH2O2
46.025
MFCD00003297
BDAGIHXWWSANSR-UHFFFAOYSA-N
methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt
284
CHEBI:30751
Ameisensäure
C(=O)O

Used as a hydrogen source, as a reducing agent and is used in tanning and dye fixing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Formic Acid
Namenshinweis For analysis ACS
Schmelzpunkt 7°bis 9 °C
Siedepunkt 100°bis 101 °C
Prozentgehaltsbereich >96+%
Strukturformel HCOOH
UN-Nummer UN1779
Beilstein 1209246
Merck Index 14,4241
Löslichkeitsinformationen Miscible with water. Partially miscible in toluene,xylene and benzene.
Formelmasse 46.03
Reinheit (%) ≥96%
Güte ACS
Dichte 1,220 g/mL
Menge 500g
Flammpunkt 50 °C (156 °F)
Physikalische Form Flüssig
Brechungsindex 1,371
Empfindlichkeit Hygroskopisch
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'