Learn More
FOXA2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen 720061
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
These Polyclonal antibodies are of rabbit origin developed by immunizing animals with proteins or peptides. The polyclonal antibody is purified by affinity purification from the rabbit sera generated after immunizing the rabbits with a specific type of protein or peptide. The purified antibody is tested for its functionality in various relevant research applications. The antibody is developed for Research Use Only and is non-hazardous or non-infectious in nature.
FOXA2 Polyclonal antibody specifically detects FOXA2 in Human samples. It is validated for Immunocytochemistry, Western Blot.Spezifikation
FOXA2 | |
Polyclonal | |
Unconjugated | |
FOXA2 | |
FKHL20; forkhead box A2; forkhead box protein A2; Foxa2; hepatic nuclear factor-3-beta; hepatocyte nuclear factor 3 beta (winged helix transcription factor); hepatocyte nuclear factor 3, beta; hepatocyte nuclear factor 3-beta; hepatocyte nuclear factor-3beta; HNF3B; Hnf-3b; HNF3beta; HNF-3BETA; HNF-3-beta; HNF3-beta; MGC19807; putative; RONU; TCF3B; TCF-3B; transcription factor; transcription factor 3B; transcription factor Foxa2; WHN | |
Rabbit | |
Affinity chromatography | |
RUO | |
3170 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 0.1% BSA, 30% glycerol and 0.09% sodium azide; pH 7.2 | |
Q9Y261 | |
FOXA2 | |
Recombinant protein corresponding to Human FOXA2 (aa 236-451) | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |