missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXI3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA548217
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
FOXI3 Polyclonal antibody specifically detects FOXI3 in Human samples. It is validated for Flow Cytometry, Western Blot.Spezifikation
FOXI3 | |
Polyclonal | |
Unconjugated | |
FOXI3 | |
forkhead box I3; forkhead box protein I3; Foxi3 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
344167 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Western Blot | |
0.45 mg/mL | |
PBS with 0.09% sodium azide | |
A8MTJ6 | |
FOXI3 | |
KLH conjugated synthetic peptide between 157-186 amino acids from the Central region of human FOXI3. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |