Learn More
FOXP2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen 720031
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
These Polyclonal antibodies are of rabbit origin developed by immunizing animals with proteins or peptides. The polyclonal antibody is purified by affinity purification from the rabbit sera generated after immunizing the rabbits with a specific type of protein or peptide. The purified antibody is tested for its functionality in various relevant research applications. The antibody is developed for Research Use Only and is non-hazardous or non-infectious in nature. This antibody is predicted to react with Monkey, Mouse, Pig and Rat.
FOXP2 Polyclonal antibody specifically detects FOXP2 in Human samples. It is validated for Immunocytochemistry, Western Blot.Spezifikation
FOXP2 | |
Polyclonal | |
Unconjugated | |
FOXP2 | |
2810043D05Rik; AI449000; CAG repeat protein 44; CAG-16; CAGH44; D0Kist7; DKFZp686H1726; forkhead box P2; forkhead box protein P2; forkhead/winged-helix transcription factor; FOXP2; OTTHUMP00000196932; RGD1559697; SPCH1; TNRC10; trinucleotide repeat containing 10; trinucleotide repeat-containing gene 10 protein | |
Rabbit | |
Affinity chromatography | |
RUO | |
93986 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 0.1% BSA, 30% glycerol and 0.09% sodium azide; pH 7.2 | |
O15409 | |
FOXP2 | |
Peptides corresponding to Human FOXP2 (aa 576-587, 670-684, 700-715). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |