Learn More
FRMPD3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA551844
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: MMKKEALLLI PNVLKVFLEN GQIKSFTFDG RTTVKDVMLT LQDRLSLRFI EHFALVLEYA GPEQNHKFLL LQDKQPLAYV VQRTHYHGMK CLFRISFFPK DPVELLRRDP AAFEYLYIQV ESGL Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 94%.
FRMPD3 Polyclonal antibody specifically detects FRMPD3 in Human samples. It is validated for Immunohistochemistry.Spezifikation
FRMPD3 | |
Polyclonal | |
Unconjugated | |
FRMPD3 | |
B930041G04; EG245643; FERM and PDZ domain containing 3; FERM and PDZ domain-containing protein 3; FRMPD3; KIAA1817; RGD1563753; RP5-1070B1.1; similar to novel protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84443 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5JV73 | |
FRMPD3 | |
Recombinant protein corresponding to Human FRMPD3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only