missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FSD2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 25609-1-AP-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
This gene encodes a protein that belongs to the FN3/SPRY family of proteins. Alternate splicing results in multiple transcript variants.Spezifikation
FSD2 | |
Polyclonal | |
Unconjugated | |
FSD2 | |
FSD2, RP11 127F21, SPRYD1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
123722, 244091, 308779 | |
-20°C | |
Liquid |
Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
A1L4K1, Q8BZ52 | |
Fsd2 | |
FSD2 Fusion Protein Ag22229 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |