missing translation for 'onlineSavingsMsg'
Learn More

Furo-[3,2-b]-pyridin-2-boronsäurepinakolester, 97 %, Thermo Scientific™

Catalog Number p-7032447
Click to view available options
Menge:
250 mg
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
2019997-40-9
C13H16BNO3
245.09
MFCD18255044
NCPRRWXGBQZDIU-UHFFFAOYSA-N
furo 3,2-b pyridine-2-boronic acid pinacol ester, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl furo 3,2-b pyridine
99738269
2-(4,4,5,5-Tetramethyl-1,3,2-Dioxaborolan-2-yl)Furo[3,2-b]Pyridin
CC1(C)OB(OC1(C)C)C1=CC2=NC=CC=C2O1
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
2019997-40-9
C13H16BNO3
245.09
MFCD18255044
NCPRRWXGBQZDIU-UHFFFAOYSA-N
furo 3,2-b pyridine-2-boronic acid pinacol ester, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl furo 3,2-b pyridine
99738269
2-(4,4,5,5-Tetramethyl-1,3,2-Dioxaborolan-2-yl)Furo[3,2-b]Pyridin
CC1(C)OB(OC1(C)C)C1=CC2=NC=CC=C2O1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material Furo[3,2-b]pyridine-2-boronic acid pinacol ester
Schmelzpunkt 163°C to 164°C
Menge 250 mg
Formelmasse 245.09
Reinheit (%) 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'