missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FXYD7 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA555463
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: KKVKCRKADS RSESPTCKSC KSELPSSAPG GGGV Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
The specific function of this protein remains unknown.This reference sequence was derived from multiple replicate ESTs and validated by similar human genomic sequence. This gene encodes a member of a family of small membrane proteins that share a 35-amino acid signature sequence domain, beginning with the sequence PFXYD and containing 7 invariant and 6 highly conserved amino acids. The approved human gene nomenclature for the family is FXYD-domain containing ion transport regulator. Transmembrane topology has been established for two family members (FXYD1 and FXYD2), with the N-terminus extracellular and the C-terminus on the cytoplasmic side of the membrane. FXYD2, also known as the gamma subunit of the Na, K-ATPase, regulates the properties of that enzyme. FXYD1 (phospholemman), FXYD2 (gamma), FXYD3 (MAT-8), FXYD4 (CHIF), and FXYD5 (RIC) have been shown to induce channel activity in experimental expression systems. This gene product, FXYD7, is novel and has not been characterized as a protein. [RefSeq curation by Kathleen J. Sweadner, Ph.D., sweadner@helix.mgh.harvard.edu.].Spezifikation
FXYD7 | |
Polyclonal | |
Unconjugated | |
FXYD7 | |
FXYD domain containing ion transport regulator 7, FLJ25096 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
53822 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P58549 | |
FXYD7 | |
Recombit protein corresponding to Human FXYD7 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only