missing translation for 'onlineSavingsMsg'
Learn More
Learn More
G6PC Antibody (CL5817) [DyLight 550], Novus Biologicals Biologicals™
Alle ansehen Bio Techne ProdukteBeschreibung
G6PC Monoclonal antibody specifically detects G6PC in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | G6PC |
| Anwendungen | Immunohistochemistry (Paraffin) |
| Klassifikation | Monoclonal |
| Klon | CL5817 |
| Konjugat | DyLight 550 |
| Zusammensetzung | 50mM Sodium Borate |
| Gen-Alias | EC 3.1.3.9, G6Pase, G-6-Pase, G6Pase-alpha, G6PT, glucose-6-phosphatase, Glucose-6-phosphatase alpha, glucose-6-phosphatase, catalytic (glycogen storage disease type I, von Gierkedisease), glucose-6-phosphatase, catalytic subunit, GSD1, GSD1a, MGC163350 |
| Wirtsspezies | Mouse |
| Immunogen | This antibody was generated using a synthetic peptide of P35575, with the exact immunogen sequence remaining proprietary. |
| Reinigungsverfahren | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?