missing translation for 'onlineSavingsMsg'
Learn More

GABRD Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marke:  Proteintech 15623-1-AP-150UL

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16884874

  • 329.85 € / 150 Mikroliter

Verfügbar ab
16-06-2021

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

GAD-65 and GAD-67, glutamate decarboxylases, function to catalyze the production of GABA (gamma-aminobutyric acid). In the central nervous system GABA functions as the main inhibitory transmitter by increasing a Cl- conductance that inhibits neuronal firing. GABA has been shown to activate both ionotropic (GABAA) and metabotropic (GABAB) receptors as well as a third class of receptors called GABAC. Both GABAA and GABAC are ligand-gated ion channels, however, they are structurally and functionally distinct. Members of the GABAA receptor family include GABAA Ralpha1-6, GABAA R beta1-3, GABAA Rgamma1-3, GABAA Rdelta, GABAA Repsilon, GABAA Rrho1 and GABAA Rrho2. The GABAB family is composed of GABAB R1alpha and GABAB R1beta. GABA transporters have also been identified and include GABA T-1, GABA T-2 and GABA T-3 (also designated GAT-1, -2, and -3). The GABA transporters function to terminate GABA action.
Spezifikation

Spezifikation

GABRD
0.24 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O14764, P18506, P22933
GABAA receptor delta, GABA A receptor delta, GABA(A) receptor subunit delta, GABRD
Rabbit
IgG
150 μL
-20°C
Primary
14403, 2563, 29689
Western Blot
Unconjugated
GABRD
Liquid
Gabrd
GABRD Fusion Protein Ag8064
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation