missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GAL3ST3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 24851-1-AP-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
GAL3ST3 is a member of the galactose-3-O-sulfotransferase protein family. It catalyzes sulfonation by transferring a sulfate group to the 3′ position of galactose in N-acetyllactosamine in both type 2 (Gal-beta-1-4GlcNAc-R) oligosaccharides and core-2-branched O-glycans, but not on type 1 or core-1-branched structures. This gene, which has also been referred to as GAL3ST2, is different from the GAL3ST2 gene located on chromosome 2 that encodes a related enzyme with distinct tissue distribution and substrate specificities, compared to galactose-3-O-sulfotransferase 3.Spezifikation
GAL3ST3 | |
Polyclonal | |
Unconjugated | |
GAL3ST3 | |
GAL3ST 3, GAL3ST2, GAL3ST3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
499302, 545276, 89792 | |
-20°C | |
Liquid |
Western Blot | |
0.46 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P61315, Q96A11 | |
Gal3st3 | |
GAL3ST3 Fusion Protein Ag20692 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |