missing translation for 'onlineSavingsMsg'
Learn More

GALNT9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marke:  Proteintech 25967-1-AP-20UL

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16884495

  • 140.00 € / 20 Mikroliter
Verfügbar ab: 18-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

This gene encodes a member of the UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase (GalNAc-T) family of enzymes. GalNAc-Ts initiate mucin-type O-linked glycosylation in the Golgi apparatus by catalyzing the transfer of GalNAc to serine and threonine residues on target proteins. They are characterized by an N-terminal transmembrane domain, a stem region, a lumenal catalytic domain containing a GT1 motif and Gal/GalNAc transferase motif, and a C-terminal ricin/lectin-like domain. GalNAc-Ts have different, but overlapping, substrate specificities and patterns of expression. This gene is expressed specifically in the brain, with highest expression in the cerebellum. Multiple transcript variants encoding different isoforms have been found for this gene.
Spezifikation

Spezifikation

GALNT9
Polyclonal
Unconjugated
GALNT9
GALNAC T9, GALNT9, pp GaNTase 9
Rabbit
Antigen Affinity Chromatography
RUO
231605, 304571, 50614
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.23 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9HCQ5
GALNT9
GALNT9 Fusion Protein Ag23233
20 μL
Primary
Human, Rat, Mouse
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote