missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GATA6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 55435-1-AP-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
GATA6 is thought to be important for regulating terminal differentiation and/or proliferation.Spezifikation
GATA6 | |
Polyclonal | |
Unconjugated | |
GATA6 | |
GATA binding factor 6, GATA binding protein 6, GATA6, Transcription factor GATA 6 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14465, 2627, 29300 | |
-20°C | |
Liquid |
Western Blot | |
0.35 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P46153, Q61169, Q92908 | |
Gata6 | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |