Learn More
GCC1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA553988
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: LELRLEETRE ALAGRAYAAE QMEGFELQTK QLTREVEELK SELQAIRDEK NQPDPRLQEL QEEAARLKSH FQAQLQQEMR Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 93%.
GCC1 Polyclonal antibody specifically detects GCC1 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
GCC1 | |
Polyclonal | |
Unconjugated | |
GCC1 | |
4932417P04Rik; Gcc1; GCC1P; GCC88; golgi coiled coil 1; Golgi coiled coil protein 1; golgi coiled-coil 1; golgi coiled-coil protein 1; GRIP and coiled-coil domain containing 1; GRIP and coiled-coil domain-containing protein 1; peripheral membrane Golgi protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79571 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.8 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96CN9 | |
GCC1 | |
Recombinant protein corresponding to Human GCC1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only