Learn More
GCN1L1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA555064
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: LLPLLIQTVE KAASQSTQVP TITEGVAAAL LLLKLSVADS QAEAKLSSFW QLIVDEKKQV FTSEKFLVMA SEDALCTVLH LTERLFLDHP HRLTGNKVQQ YHRALVAVLL SRTWHVRRQA QQTVRKLLSS L Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 87%.
GCN1L1 Polyclonal antibody specifically detects GCN1L1 in Human samples. It is validated for Immunohistochemistry.Spezifikation
GCN1L1 | |
Polyclonal | |
Unconjugated | |
GCN1 | |
4932409G22; AL022764; eIF-2-alpha kinase activator GCN1; G431004K08Rik; Gcn1; GCN1 (general control of amino-acid synthesis 1, yeast)-like 1; GCN1 eIF2 alpha kinase activator homolog; GCN1 eIF-2-alpha kinase activator homolog; GCN1 general control of amino-acid synthesis 1-like 1; GCN1 general control of amino-acid synthesis 1-like 1 (yeast); GCN1, eIF2 alpha kinase activator homolog; GCN1L; GCN1L1; GCN1-like protein 1; General control of amino-acid synthesis 1-like protein 1; hsGCN1; KIAA0219; mKIAA0219; peroxisome proliferator activated receptor interacting complex protein; PRIC295; translational activator GCN1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10985 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q92616 | |
GCN1 | |
Recombinant protein corresponding to Human GCN1L1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only