missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GCP6 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 25022-1-AP-20UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
The protein encoded by this gene is part of a large multisubunit complex required for microtubule nucleation at the centrosome.Spezifikation
GCP6 | |
Polyclonal | |
Unconjugated | |
TUBGCP6 | |
GCP 6, GCP6, KIAA1669, TUBGCP6 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
85378 | |
-20°C | |
Liquid |
Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q96RT7 | |
TUBGCP6 | |
GCP6 Fusion Protein Ag21634 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |