Learn More
GDA Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA556455
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: FRGTFVHSTW TCPMEVLRDH LLGVSDSGKI VFLEEASQQE KLAKEWCFKP CEIRELSHHE FFMPGLVDTH IHASQYSFAG SSIDLPLLEW LT Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 92%.
GDA Polyclonal antibody specifically detects GDA in Human samples. It is validated for Immunohistochemistry, Western Blot.Spezifikation
GDA | |
Polyclonal | |
Unconjugated | |
GDA | |
AU015411; AW047581; Cypin; cytoplasmic PSD95 interactor; GAH; Gda; GUANASE; guanine aminase; guanine aminohydrolase; Guanine deaminase; KIAA1258; NEDASIN; p51-nedasin | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9615 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y2T3 | |
GDA | |
Recombinant protein corresponding to Human GDA | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only