Learn More
GEM Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA555188
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: NTYYRVVLIG EQGVGKSTLA NIFAGVHDSM DSDCEVLGED TYERTLMVDG ESATIILLDM WENKGENEWL HDHCMQVGDA YLIVYSITD Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 56%.
GEM Polyclonal antibody specifically detects GEM in Human samples. It is validated for Immunohistochemistry, Western Blot.Spezifikation
GEM | |
Polyclonal | |
Unconjugated | |
GEM | |
AV020497; Gem; GTP binding protein (gene overexpressed in skeletal muscle); GTP binding protein overexpressed in skeletal muscle; GTP-binding mitogen-induced T-cell protein; GTP-binding protein GEM; GTP-binding protein overexpressed in skeletal muscle; kinase-inducible Ras-like protein; KIR; MGC26294; RAS-like protein KIR | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2669 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P55040 | |
GEM | |
Recombinant protein corresponding to Human GEM | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only