missing translation for 'onlineSavingsMsg'
Learn More

Gemin 2 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Thermo Scientific PA561251

Weitere Details : Netto-Gewicht : 0.02000kg

Artikelnummer. 15739071

  • 477.00 € / 100 Mikroliter
Verfügbar ab: 23-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Immunogen sequence: QWQQQQVAQF STVRQNVNKH RSHWKSQQLD SNVTMPKSED EEGWKKFCLG EKLCADGAVG PATNESPGID YVQIGFPPLL SIVSRMNQ Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 88%.

Gemin 2 Polyclonal antibody specifically detects Gemin 2 in Human samples. It is validated for Immunohistochemistry.
Spezifikation

Spezifikation

Gemin 2
Polyclonal
Unconjugated
GEMIN2
1700012N19Rik; Component of gems 2; gem (nuclear organelle) associated protein 2; gem nuclear organelle associated protein 2; gem nuclear organelle associated protein 2 S homeolog; gem-associated protein 2; GEMIN2; Gemin-2; gemin2.S; SIP 1; SIP1; SIP-1; SIP1 delta; sip1-delta; SMN interacting protein 1-delta; SMN interacting protein-1; SMN-interacting protein 1; survival of motor neuron protein interacting protein 1; Survival of motor neuron protein-interacting protein 1; survivor of motor neuron protein interacting protein 1; XELAEV_18042638mg
Rabbit
Antigen affinity chromatography
RUO
8487
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry
0.4 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
O14893
GEMIN2
Recombinant protein corresponding to Human Gemin 2
100 μL
Primary
Human
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote

Research Use Only