Beschreibung
Geminin Polyclonal specifically detects Geminin in Human samples. It is validated for Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Western Blot (Negative).
Spezifikation
Spezifikation
| Antigen | Geminin |
| Anwendungen | Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot |
| Klassifikation | Polyclonal |
| Konzentration | 1.0 mg/mL |
| Konjugat | Unconjugated |
| Verdünnung | Immunohistochemistry 1:10-1:500, Immunoprecipitation 2-5 ug/mg lysate, Immunohistochemistry-Paraffin 1:2000 - 1:10000, Western Blot (Negative) |
| Zusammensetzung | Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide |
| Gen-Zugriffsnummer | NP_056979.1 |
| Gen-Alias | Gem, geminin, geminin, DNA replication inhibitor, RP3-369A17.3 |
| Gensymbole | GMNN |
| Mehr anzeigen |