missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GEN1 Recombinant Rabbit Monoclonal Antibody (K01_1R62)
GREENER_CHOICE

Artikelnummer. p-200113955
Change view
Click to view available options
Menge:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge
30826219 100 μL
30828163 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 30826219 Lieferant Invitrogen™ Lieferanten-Nr. MA563453

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Rabbit Recombinant Monoclonal Antibody

GEN1 Recombinant Monoclonal Antibody for Western Blot

Xeroderma pigmentosum type G (XPG) is a human genetic disease exhibiting extreme sensitivity to sunlight. The XPG protein, a member of the flap endonuclease 1 (FEN-1) structure-specific DNA repair endonuclease family, is an enzyme essential for DNA repair of the major kinds of solar ultraviolet (UV)-induced DNA damages. Human XPG nuclease makes the 3' incision during nucleotide excision repair of DNA. The enzyme cleaves model DNA bubble structures specifically near the junction of unpaired DNA with a duplex region. A 29-amino acid region of human XPG (residues 981-1009) contains the PCNA binding activity. A conserved arginine in XPG (Arg992) is crucial for its PCNA binding activity. Replication Protein A (RPA) binds specifically and directly to two excision repair proteins, the xeroderma pigmentosum damage-recognition protein XPA and the endonuclease XPG.
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen GEN1
Anwendungen Western Blot
Klassifikation Recombinant Monoclonal
Klon K01_1R62
Konzentration 2 mg/mL
Konjugat Unconjugated
Zusammensetzung 50mM tris glycine with 40% glycerol, 150mM NaCl, 0.05% BSA and 0.01% sodium azide; pH 7.4
Gen GEN1
Gen-Zugriffsnummer Q17RS7, Q8BMI4
Gen-Alias 5830483C08Rik; Flap endonuclease GEN homolog 1; Gen; Gen endonuclease homolog 1; Gen homolog 1, endonuclease; Gen homolog 1, endonuclease (Drosophila); GEN1; GEN1 Holliday junction 5' flap endonuclease; GEN1, Holliday junction 5' flap endonuclease; Holliday junction resolvase; RGD1559792
Gensymbole GEN1
Wirtsspezies Rabbit
Immunogen A synthetic peptide of human GEN1.
Reinigungsverfahren Protein A/G
Menge 100 μL
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 209334, 298884, 348654
Zielspezies Human, Mouse, Rat
Inhalt und Lagerung Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Produkttyp Antibody
Form Liquid
Isotype IgG
Mehr anzeigen Weniger anzeigen
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.