missing translation for 'onlineSavingsMsg'
Learn More

AcroBiosystems GENPowerTM AsCas12a Nuclease (active enzyme, MALS verified)

Artikelnummer. 31453544
Change view
Click to view available options
Menge:
50 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge
31453544 50 μg
31453174 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 31453544 Lieferant AcroBiosystems Lieferanten-Nr. CAAA514350ug

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Recombinant Protein

GENPowerTM AsCas12a Nuclease is expressed from Acidaminococcus sp. BV3L6 E. coli.

Spezifikation

Format Liquid
Zusammensetzung Supplied as 0.2 μm filtered solution in 20 mM Tris, 300 mM NaCl, pH 7.5 with glycerol as protectant. Contact us for customized product form or formulation.
Qualitätskontrollen Less than 0.1 EU per μg by the LAL method / rFC method.
Menge 50 μg
Spezies Acidaminococcus (BV3L6)
Endotoxin-Gehalt Less than 0.1 EU per μg by the LAL method / rFC method.
Versandbedingung This product is supplied and shipped with dry ice, please inquire the shipping cost.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.