missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GFM2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 16941-1-AP-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
GFM2 is involved in mitochondrial translation, which is crucial for maintaining mitochondrial function and mutations in this system lead to a breakdown in the respiratory chain-oxidative phosphorylation system and to impaired maintece of mitochondrial DNA. In Eukaryotes contain two protein translational systems, one in the cytoplasm and one in the mitochondria. This gene encodes one of mitochondrial translation elongation factors. Its role in the regulation of normal mitochondrial function and in different disease states attributed to mitochondrial dysfunction is not known. Alternative splicing results in at least three transcript variants encoding distinct isoforms.Spezifikation
GFM2 | |
Polyclonal | |
Unconjugated | |
GFM2 | |
EF G2mt, EFG2, Elongation factor G2, GFM2, hEFG2, mEF G 2, MST027, RRF2mt | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
294672, 320806, 84340 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q5BJP6, Q8R2Q4, Q969S9 | |
Gfm2 | |
GFM2 Fusion Protein Ag10530 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |