Learn More
IBA Lifesciences GFP-Strep-tag control protein
GFP-Strep-tag control protein for Strep-Tactin™ and Strep-Tactin™XT applications
Marke: IBA Lifesciences 2-1007-005
Weitere Details : Netto-Gewicht : 0.00050kg
Beschreibung
- GFP control protein consisting of the green fluorescent protein (GFP) C-terminally tagged with the Strep-tag™II or Twin-Strep-tag™.
- It is suitable for test purifications, detections, and immobilizations with Strep-Tactin™ or Strep-Tactin™XT.
Spezifikation
0.5 mL | |
500 μg of a 30 kDa protein | |
Twin-Strep-tag™ |
For Research Use Only.