Learn More
GGACT Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA564214
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: TLEPYPLVIA GEHNIPWLLH LPGSGRLVEG EVYAVDERML RFLDDFESCP Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 78%.
The protein encoded by this gene aids in the proteolytic degradation of crosslinked fibrin by breaking down isodipeptide L-gamma-glutamyl-L-epsilon-lysine, a byproduct of fibrin degradation. The reaction catalyzed by the encoded gamma-glutamylaminecyclotransferase produces 5-oxo-L-proline and a free alkylamine. Two transcript variants encoding the same protein have been found for this gene.Spezifikation
GGACT | |
Polyclonal | |
Unconjugated | |
GGACT | |
A2ld1; AIG2-like domain 1; AIG2-like domain-containing protein 1; Gamma-glutamylamine cyclotransferase; gamma-glutamylaminecyclotransferase; GGACT; RGD1304748 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
87769 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BVM4 | |
GGACT | |
Recombinant protein corresponding to Human GGACT | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only