Learn More
GGT6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA555546
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: EEPVVYQKLL PWEPSLESEE EVEEEETSEA LVLNPRRHQD SSRNKAGGLP GTWVRVVAAL LLLAVGCSLA VRQLQNQGRS Highest antigen sequence identity to the following orthologs: Mouse - 56%, Rat - 59%.
GGT6 Polyclonal antibody specifically detects GGT6 in Human samples. It is validated for Immunohistochemistry, Western Blot.Spezifikation
GGT6 | |
Polyclonal | |
Unconjugated | |
GGT6 | |
9030405D14Rik; AI427679; AW490214; gamma-glutamyl transferase 6; gamma-glutamyltransferase 6; gamma-glutamyltransferase 6 homolog; gamma-glutamyltranspeptidase 6; GGT 6; GGT6; Glutathione hydrolase 6; Glutathione hydrolase 6 heavy chain; Glutathione hydrolase 6 light chain | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
124975 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6P531 | |
GGT6 | |
Recombinant protein corresponding to Human GGT6 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only