Learn More
GGT7 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA553035
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: HLVLSPPPPH TGPALISALN ILEGFNLTSL VSREQALHWV AETLKIALAL ASRLGDPVYD STITESMDDM LSKVEAAYLR GHINDSQAAP APLLPVYELD GAPTAAQVLI MGPDDFIVAM VSSLNQPFGS GLITPSGILL NS Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 95%.
GGT7 Polyclonal antibody specifically detects GGT7 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Western Blot.Spezifikation
GGT7 | |
Polyclonal | |
Unconjugated | |
Ggt7 | |
1110017C11Rik; 6330563L03Rik; D20S101; dJ18C9.2; gamma-glutamyltransferase 4; gamma-glutamyltransferase 7; gamma-glutamyltransferase 7; glutathione hydrolase 7; gamma-glutamyltransferase-like 3; Gamma-glutamyltransferase-like 5; gamma-glutamyltranspeptidase 7; gamma-glutamyltranspeptidase-like 3; GGT 7; GGT4; Ggt7; GGTL3; GGTL5; Glutathione hydrolase 7; Glutathione hydrolase 7 heavy chain; Glutathione hydrolase 7 light chain; LOC100732094; mFLJ00311 protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
156275, 207182, 2686 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q99JP7, Q99MZ4, Q9UJ14 | |
Ggt7 | |
Recombinant protein corresponding to Human GGT7 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Research Use Only