Learn More
Gibco™ Human beta-Defensin 1 Recombinant Protein
Human beta-Defensin 1 Recombinant Protein
Marke: Gibco™ PHC1624
Weitere Details : Netto-Gewicht : 0.50000kg
Beschreibung
Human beta-Defensin 1 Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN
BD1 is a member of the defensin family involved in defense against mibcrobes.Approved for shipment at Room Temperature or on Wet or Dry Ice.
Spezifikation
Cell Culture | |
3.9 kDa | |
20 ug | |
36 aa | |
Store in freezer (-5 to -30°C). | |
< 0.1 ng/μg | |
Approved for shipment on Wet or Dry Ice | |
Cytokines & Receptors | |
Lyophilized | |
Immunology | |
98 % |
1672 | |
Sequential Chromatography | |
BD-1 | |
Human | |
Carrier-Free | |
20 μg | |
E. coli | |
Recombinant, Ligand | |
Other Proteins | |
Gibco™ |
For Research Use Only. Not for use in diagnostic procedures.